missing translation for 'onlineSavingsMsg'
Learn More
Learn More
First Aid Kit Plasters
Description
ul>Various sizes, colors and styles Included: Washproof plasters, fabric plasters and detectable blue plasters for catering industry Strong, durable and highly conformable
Specifications
Specifications
| Material | Fabric |
| Product Type | HypaPlast Pink Plaster |
| Sterility | Sterile |
| Quantity | 100 Pk. |
| Size | Assorted |
| Packaging Type | Individually Wrapped |
| Waterproof | Yes |
| For Use With (Application) | Multi-purpose, First aid kit |
| Color | Pink |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?