missing translation for 'onlineSavingsMsg'
Learn More

First Aid Kit Plasters

Product Code. 12960455
Change view
Click to view available options
Product Type:
HypaPlast Blue Catering Plaster
HypaPlast Pink Plaster
HypaPlast Plaster
Size:
7.2 x 2.5 cm
Assorted
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Size unitSize
12960455 HypaPlast Blue Catering Plaster Assorted Each
12950455 HypaPlast Plaster 7.2 x 2.5 cm Each
12930455 HypaPlast Plaster Assorted Each
12940455 HypaPlast Pink Plaster 7.2 x 2.5 cm Each
12920455 HypaPlast Pink Plaster Assorted Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12960455 Supplier Safety First Supplier No. D7010
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use First Aid Kit Plasters as an integral part of your first aid kit supplies for workplace, school, or home.

ul>
  • Various sizes, colors and styles
  • Included: Washproof plasters, fabric plasters and detectable blue plasters for catering industry
  • Strong, durable and highly conformable
  • Specifications

    Material Fabric
    Product Type HypaPlast Blue Catering Plaster
    Sterility Sterile
    Quantity 100 Pk.
    Size Assorted
    Packaging Type Individually Wrapped
    Waterproof Yes
    For Use With (Application) Catering and food handling
    Color Blue
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.