Learn More
Safety First Aid HypaSoothe Emergency Burn Gel, Small
Product Code. 31759451
Change view
Click to view available options
Size:
Small
Unit Size:
Pack of 20
Product Code. Size Unit Size
31759451 Small Pack of 20
1 options
Product Code. 31759451 Supplier Safety First Aid Supplier No. D8163PK20
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Gel is a sterile wound hydrogel that facilitates moist wound healing for minor burns. The gel re-hydrates necrotic tissue while loosening and absorbing slough and exudate. The gel is suitable for use on 1st and 2nd degree burns.
  • The gel is suitable for use on 1st and 2nd degree burns.
  • It soothes burns and reduces the likelihood of scarring by creating an ideal wound healing environment.
  • High cohesion means the gel is easily removed, even after the absorption of debris and exudate.
  • Supplied in a 20 pack of 6 g sachets.
  • Usage: Cool the burn with water. Gently dry the skin around the wound and apply the gel Cover the wound with a secondary dressing. Caution: For external use only. Do not reuse gel that has already been applied. Do not use if packaging is broken.
  • In the case of an allergic reaction, stop usage and consult a healthcare professional
Specifications
Material Cloth
Product Type Burn Dressing
Sterility Non-sterile
Quantity 6 g
Size Small
Packaging Type Sachets