Learn More
Thermo Scientific™ HyPerforma™ Smartainer™ 3.0 Bioprocess Container (BPC), Aegis™ 5-14 film
GREENER_CHOICE
Product Code. 17978261 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
200 L
500 L
Insert Type:
Horizontally-gusseted
Vertically-gusseted
Unit Size:
Each
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Insert Type unitSize
17978261 500 L Vertically-gusseted Pack of 5
17958221 200 L Horizontally-gusseted Pack of 5
17948221 500 L Horizontally-gusseted Pack of 5
31940830 500 L Horizontally-gusseted Each
31940834 500 L Vertically-gusseted Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17978261 Supplier Thermo Scientific™ Supplier No. SH31188.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The Thermo Scientific HyPerforma Smartainer 3.0 BPC provides end-users producing biologics or vaccine therapeutics with a large-volume, sterile liquid storage system for volumes ranging from 200 L to 3000 L—designed for the collection and storage of sterile process liquids such as cell culture media, process water or purification media. Options include temperature-control through integrated water jackets, BPC deploying hoists, as well as other options to optimize performance in your productions processes.
The Thermo Scientific HyPerforma Smartainer 3.0 BPC provides end-users producing biologics or vaccine therapeutics with a large-volume, sterile liquid storage system for volumes ranging from 200 L to 3000 L—designed for the collection and storage of sterile process liquids such as cell culture media, process water or purification media. Options include temperature-control through integrated water jackets, BPC deploying hoists, as well as other options to optimize performance in your productions processes.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 500 L
Product Type Bioprocess Container (BPC)
Insert Type Vertically-gusseted