missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyperSep™ Dispersive SPE Clean-up Products (QuEChERS)

Product Code. 10649275 Shop All Thermo Scientific Products
Click to view available options
Volume (Metric):
15 mL
15 mL Centrifuge Tubes
2 mL Centrifuge Tubes
6 mL Centrifuge Tubes
For Use With (Application):
QuEChERS Method
Schenck Method
Description:
1200 mg MgSO4, 400 mg PSA
1200 mg MgSO4, 400 mg PSA, 400 mg C18
1200 mg MgSO4, 400 mg PSA, 400 mg C18, 400 mg GCB
1200 mg MgSO4, 900 mg PSA
150 mg MgSO4, 150 mg PSA
150 mg MgSO4, 150 mg PSA, 50 mg C18
150 mg MgSO4, 25 mg PSA
150 mg MgSO4, 25 mg PSA, 2.5 mg GCB
150 mg MgSO4, 25 mg PSA, 25 mg C18
150 mg MgSO4, 25 mg PSA, 7.5 mg GCB
150 mg MgSO4, 300 mg PSA, 150 mg Chlorofiltr
150 mg MgSO4, 50 mg PSA
150 mg MgSO4, 50 mg PSA, 50 mg C18
150 mg MgSO4, 50 mg PSA, 50 mg C18, 50 mg GCB
150 mg MgSO4, 50 mg PSA, 50 mg Chlorofiltr
150 mg MgSO4, 50 mg PSA, 50 mg GCB
400 mg PSA on Bottom, 200 mg GCB on Top, Separated by a Frit
4000 mg MgSO4, 1000 mg NaCl, 1000 mg C6H9Na3O9, 500 mg C6H6Na2O7 · 1.5 H2O
500 mg PSA on Bottom, 250 mg GCB on Top, Separated by a Frit
500 mg PSA on Bottom, 500 mg GCB on Top, Separated by a Frit
750 mg MgSO4, 250 mg PSA, 250 mg C18, 250 GCB
900 mg MgSO4, 150 mg PSA
900 mg MgSO4, 150 mg PSA, 15 mg GCB
900 mg MgSO4, 150 mg PSA, 150 mg C18
900 mg MgSO4, 150 mg PSA, 45 mg GCB
900 mg MgSO4, 300 mg PSA
900 mg MgSO4, 300 mg PSA, 150 mg C18
900 mg MgSO4, 300 mg PSA, 150 mg GCB
900 mg anhydrous magnesium sulfate, 150 mg PSA
900 mg anhydrous magnesium sulfate, 300 mg PSA
Unit Size:
Pack of 10
Pack of 100
Pack of 30
Pack of 50
Pack of 500
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 10649275

Marca: Thermo Scientific™ 60105230

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Save time and achieve more accurate pesticide determinations using these pre-prepared dispersive SPE products for QuEChERS clean-up.

QuEChERS Dispersive SPE methods offer a convenient and effective approach for extraction and clean-up of pesticide residues in food and other complex matrices. The robust procedure offers a number of compelling advantages: high recoveries, accurate results, high sample throughput, low solvent and glassware usage, reduced labor and bench space and lower reagent costs.

Thermo Scientific™ HyperSep Dispersive SPE Clean-Up Products are available in a range of pre-prepared sorbent combinations, each containing the proper sorbents for optimum clean-up of analytes extracted using QuEChERS methods.

HyperSep Dispersive SPE Clean-Up Products contain PSA (primary/secondary amine) for the removal of organic acids and polar pigments among other compounds. Some products couple the PSA with endcapped C18 for the removal of most lipids and sterols, or graphitized carbon black for the removal of sterols and pigments such as chlorophyll.

(A variety of tube formats and bed weights are available to accommodate large and small sample sizes.)

All HyperSep Dispersive SPE products are pre-prepared and optimized for the method at hand, so you will:

  • Save valuable preparation time
  • Achieve more consistent analytical results due to consistency in products and rigorous quality control
  • Get cleaner extracts and lower peak counts as a result of using highest quality, pre-prepared products

QuEChERS Technique

Developed by the USDA Eastern Regional Research Center in 2003, the QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) technique is increasingly becoming the technique of choice for extraction and clean-up of pesticide residues.

The QuEChERS method is a two-step process: extraction followed by clean-up. There are four variations of the QuEChERS method currently in use:

  • The original QuEChERS method uses sodium chloride to enhance extraction
  • Dispersive AOAC 2007.01 uses sodium acetate as a buffer, replacing sodium chloride
  • The Dual-Phase variation introduces PSA and GCB to remove high levels of chlorophyll and plant sterols in the final extract without the loss of planar pesticides (polar aromatics) using an acetone:toluene solvent mix (3:1)
  • European Version is similar to the AOAC method, except the extraction uses sodium chloride, sodium citrate dihydrate and disodium citrate sesquihydrate instead of sodium acetate
TRUSTED_SUSTAINABILITY

Specifica

Description 150 mg MgSO4, 50 mg PSA, 50 mg GCB
For Use With (Application) QuEChERS Method
Product Line HyperSep
Sorbent Anhydrous Magnesium Sulfate, Primary/Secondary Amine, Graphitized Carbon Black
Type QuEChERS Clean-up
Volume (Metric) 15 mL Centrifuge Tubes

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato