Description
Use the Thermo Scientific™ HyperSep™ Positive Pressure Manifold Waste Container to collect waste from the manifold. The HyperSep Positive Pressure Manifold holds up to 48 SPE cartridges. Samples and solvent, added to the SPE cartridge, are extracted under positive pressure for an even flow at each position. Extracts are collected into 13 or 16mm tubes.
Specifications
Specifications
| Description | Pre-Drilled Waste Container, 1 pk |
| For Use With (Application) | SPE |
| For Use With (Equipment) | Positive Pressure Manifold |
| Product Line | HyperSep |
| Type | Pre-Drilled Waste Container |