Learn More
Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Extraction Tips, 1-10μL
Product Code. 10364513 Shop All Thermo Scientific Products
Change view
Click to view available options
Sorbent:
BioBasic C18
BioBasic C4
BioBasic C8
HyperSep HILIC
HyperSep Hypercarb
HyperSep Hypercarb and C18 (Mix Mode)
HyperSep Poros SAX
HyperSep Poros SCX
HyperSep Poros WAX
HyperSep Titanium Dioxide
HyperSep Trypsin
HyperSep Zirconium Dioxide
Description:
1 to 10 μL Volume
10 to 200 μL Volume
Unit Size:
Pack of 96
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sorbent Description unitSize
10364513 BioBasic C18 1 to 10 μL Volume Pack of 96
10488684 BioBasic C8 1 to 10 μL Volume Pack of 96
10662094 BioBasic C4 1 to 10 μL Volume Pack of 96
10314043 HyperSep Hypercarb 1 to 10 μL Volume Pack of 96
10522075 HyperSep Hypercarb and C18 (Mix Mode) 1 to 10 μL Volume Pack of 96
10304673 HyperSep HILIC 1 to 10 μL Volume Pack of 96
10611016 HyperSep Trypsin 1 to 10 μL Volume Pack of 96
10610046 HyperSep Poros WAX 1 to 10 μL Volume Pack of 96
10620046 HyperSep Poros SAX 1 to 10 μL Volume Pack of 96
10534085 HyperSep Poros SCX 1 to 10 μL Volume Pack of 96
10522265 HyperSep Titanium Dioxide 1 to 10 μL Volume Pack of 96
10488874 BioBasic C8 10 to 200 μL Volume Pack of 96
10706334 BioBasic C4 10 to 200 μL Volume Pack of 96
10065694 HyperSep Zirconium Dioxide 1 to 10 μL Volume Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 10364513 Supplier Thermo Scientific™ Supplier No. 60109401
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Get faster sample preparation with minimal sample loss with these SPE micropipette tips and achieve separation in volumes as low as 100nL.
Get faster sample preparation with minimal sample loss with the revolutionary Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Tips and achieve separation in volumes as low as 100nL.
Features:
Ideal for small sample volumes; features a 1 to 2μm wide slit at the bottom that permits liquid to pass through while retaining the chromatographic material (20 to 30μm).
  • [bullet] Filter-free design to eliminate dead volume
  • [bullet] No contamination from the supporting matrix
  • [bullet] Available in many popular chemistries
Applications include:
  • [bullet] Mass spectrometry
  • [bullet] Desalting
  • [bullet] Protein purification
  • [bullet] MALDI
  • [bullet] Electrophoresis
  • [bullet] HPCE
  • [bullet] HPLC
  • [bullet] CEC
TRUSTED_SUSTAINABILITY
Specifications
Description 1 to 10 μL Volume
For Use With (Application) Microscale Solid Phase Extraction
For Use With (Equipment) Micropipettes
Particle Size 40 to 60 μm
Product Line HyperSep
Sorbent BioBasic C18
Type SpinTip
Volume (Metric) 1 to 10 μL
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.