Learn More
Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Extraction Tips, 10-200μL
Product Code. 10111273 Shop All Thermo Scientific Products
Change view
Click to view available options
Sorbent:
BioBasic C18
HyperSep HILIC
HyperSep Hypercarb
HyperSep Hypercarb and C18 (Mix Mode)
HyperSep Poros SAX
HyperSep Poros SCX
HyperSep Poros WAX
HyperSep Titanium Dioxide
HyperSep Trypsin
HyperSep Zirconium Dioxide
Unit Size:
Pack of 96
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sorbent unitSize
10111273 HyperSep Hypercarb Pack of 96
10404603 BioBasic C18 Pack of 96
10535435 HyperSep Zirconium Dioxide Pack of 96
10140503 HyperSep Titanium Dioxide Pack of 96
10138004 HyperSep Poros SCX Pack of 96
10498684 HyperSep Poros SAX Pack of 96
10341145 HyperSep Poros WAX Pack of 96
10426744 HyperSep Trypsin Pack of 96
10314673 HyperSep HILIC Pack of 96
10716334 HyperSep Hypercarb and C18 (Mix Mode) Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 10111273 Supplier Thermo Scientific™ Supplier No. 60109415
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Get faster sample preparation with minimal sample loss with these SPE micropipette tips and achieve separation in volumes as low as 100nL.
Get faster sample preparation with minimal sample loss with the revolutionary Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Tips and achieve separation in volumes as low as 100nL.
Features:
Ideal for small sample volumes; features a 1 to 2μm wide slit at the bottom that permits liquid to pass through while retaining the chromatographic material (20 to 30μm).
  • [bullet] Filter-free design to eliminate dead volume
  • [bullet] No contamination from the supporting matrix
  • [bullet] Available in many popular chemistries
Applications include:
  • [bullet] Mass spectrometry
  • [bullet] Desalting
  • [bullet] Protein purification
  • [bullet] MALDI
  • [bullet] Electrophoresis
  • [bullet] HPCE
  • [bullet] HPLC
  • [bullet] CEC
TRUSTED_SUSTAINABILITY
Specifications
Description Hypercarb; 10-200 uL Vol.
For Use With (Application) Microscale Solid Phase Extraction
For Use With (Equipment) Micropipettes
Particle Size 30 μm
Product Line HyperSep
Sorbent HyperSep Hypercarb
Type SpinTip
Volume (Metric) 10 to 200 μL
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.