Learn More
uvex Hypervision CA FBL. AF GY/GY/BU
Product Code. 31751170 Shop All Uvex Products
Change view
Click to view available options
Lens Material:
Polycarbonate
Unit Size:
Each
Product Code. Lens Material Unit Size
31751170 Polycarbonate Each
1 options
Product Code. 31751170 Supplier uvex Supplier No. 9321614
  • ONLINE EXCLUSIVE PRICE
    17.10 €
    16.38 € / Pack of 1
    Save 0.72 €
    4% Off
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Goggles uvex hypervision Clear 9321614
  • The uvex hypervision features a comfortable, ergonomic design with a translucent frame that provides an almost unrestricted field of vision for enhanced workplace awareness.
  • A soft, flexible frame construction ensures high wearer comfort, even during extended periods of use.
  • Integrated side recesses allow sufficient space for personal prescription spectacles, enabling the uvex hypervision to be worn comfortably as reliable overspecs.
  • An adjustable headband combined with indirect ventilation completes the design, delivering a secure fit, improved airflow, and dependable eye protection for everyday professional applications.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant, Impact-resistant, Anti-fog
Lens Tint None
Product Type Goggles
Material Plastic
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection None
Frame Type Frameless
Autoclavable Not Autoclavable