Learn More
uvex Hypervision PC Foam Clear SV Exc GY/BU
Product Code. 31751168 Shop All Uvex Products
Change view
Click to view available options
Lens Material:
Polycarbonate
Unit Size:
Each
Product Code. Lens Material Unit Size
31751168 Polycarbonate Each
1 options
Product Code. 31751168 Supplier uvex Supplier No. 9321266
  • ONLINE EXCLUSIVE PRICE
    18.71 €
    17.93 € / Pack of 1
    Save 0.78 €
    4% Off
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Goggles uvex hypervision Clear sv exc. 9321266
  • The uvex hypervision features a comfortable, ergonomic design with a translucent frame that provides an almost unrestricted field of vision for enhanced workplace awareness.
  • A soft, flexible frame construction ensures high wearer comfort, even during extended periods of use.
  • Integrated side recesses allow sufficient space for personal prescription spectacles, enabling the uvex hypervision to be worn comfortably as reliable overspecs.
  • An adjustable headband combined with indirect ventilation completes the design, delivering a secure fit, improved airflow, and dependable eye protection for everyday professional applications.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant, Impact-resistant, Anti-fog
Lens Tint None
Product Type Anti-fog Goggles
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400
Frame Type Classic
Autoclavable Not Autoclavable