missing translation for 'onlineSavingsMsg'
Learn More

Hypophosphorous acid, 50% w/w aq. soln.

Product Code. 11327747
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327747 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11327747 Supplier Thermo Scientific Alfa Aesar Supplier No. 014142.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6303-21-5 | H3O2P | 66.00 g/mol

Hypophosphorous acid is primarily used for electroless nickel plating. It is involved in the reduction of arenediazonium salts. It acts as an additive in Fischer esterification reactions. Also, it serves as a neutralizing agent, antioxidant, catalyst in polymerization and poly condensation, and wetting agent. Further, it is used in the formulation of pharmaceuticals, discoloration of polymers, water treatment and retrieval of precious or non-ferrous metals. In addition to this, it is used as bleaching agents for plastics, synthetic fibers, decolorizing agent and for color stabilization during the manufacture of chemicals and several plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
  • Primarily used for electroless nickel plating.
  • Involved in the reduction of arenediazonium salts.
  • As an additive in Fischer esterification reactions.
  • Serves as a neutralizing agent, antioxidant, catalyst in polymerization and poly condensation, and wetting agent.
  • Used in the formulation of pharmaceuticals, discoloration of polymers, water treatment and retrieval of precious or non-ferrous metals.
  • Used as bleaching agents for plastics, synthetic fibers, decolorizing agent and for color stabilization during the manufacture of chemicals and several plastics
Incompatible with strong oxidizers, mercuric oxide, mercury(II) nitrate, metals and strong bases
TRUSTED_SUSTAINABILITY

Specifications

CAS 6303-21-5
Color Colorless
Density 1.225
Odor Pungent
Vapor Pressure 23 hPa (17 mm Hg)
Molecular Formula H3O2P
MDL Number MFCD02183592
UN Number UN3264
Quantity 500 g
Physical Form Liquid
Synonym hypophosphite ion, hypophosphorus acid, phosphinic acid, ion 1-, hydroxyphosphanone, oxo-, e5-phosphanol, hydroxidooxidophosphorus, perfluoroalkyl c6-c12 phosphonic acid, diperfluoroalkyl c6-c12 phosphinic acid, hydroxy-, e5-phosphanone, perfluoro-c6-c12 ethyl hydrogen phosphate
Solubility Information Miscible with water, alcohol and ether
InChI Key ACVYVLVWPXVTIT-UHFFFAOYSA-N
SMILES O[PH2]=O
IUPAC Name phosphinic acid
Molecular Weight (g/mol) 66.00
PubChem CID 22497
ChEBI CHEBI:36363
Formula Weight 66.00
Chemical Name or Material Hypophosphorous acid, 50% w/w aqueous solution
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.