missing translation for 'onlineSavingsMsg'
Learn More

Hypophosphorous acid, 50 wt.% solution in wate

Product Code. 10274252 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10274252 1 L 1 L
10680839 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10274252 Supplier Thermo Scientific Acros Supplier No. 201000010
  • ONLINE EXCLUSIVE PRICE
    228.00 €
    221.00 € / 1 L
    Save 7.00 €
    3% Off
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 6303-21-5 | H3O2P | 66.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6303-21-5 , 7732-18-5
Molecular Formula H3O2P
Molecular Weight (g/mol) 66.00
MDL Number MFCD02183592
InChI Key ACVYVLVWPXVTIT-UHFFFAOYSA-N
Synonym hypophosphite ion, hypophosphorus acid, phosphinic acid, ion 1-, hydroxyphosphanone, oxo-, e5-phosphanol, hydroxidooxidophosphorus, perfluoroalkyl c6-c12 phosphonic acid, diperfluoroalkyl c6-c12 phosphinic acid, hydroxy-, e5-phosphanone, perfluoro-c6-c12 ethyl hydrogen phosphate
PubChem CID 22497
ChEBI CHEBI:36363
IUPAC Name phosphinic acid
SMILES O[PH2]=O

Specifications

Melting Point -25.0°C
Boiling Point 108.0°C (1013.0 hPa)
Density 1.27 g/mL
Packaging Plastic Bottle
Quantity 1 L
Linear Formula H3PO2
Solubility Information Solubility in water: soluble. Other solubilities: miscible with ethanol, immiscible with hydrocarbons and halogenated, hydrocarbons
Formula Weight 66
Chemical Name or Material Hypophosphorous acid
compliance-icons
Product Identifier
  • Hypophosphorous acid, 50 wt.% solution in water
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.