missing translation for 'onlineSavingsMsg'
Learn More

Hypophosphorous Acid Solution, Honeywell

Product Code. 15661970 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit. Quantity missing translation for 'unitSize'
15661970 500 mL 500mL
15621970 100 mL 100mL
15631970 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 15661970 missing translation for 'mfr' Honeywell Fluka missing translation for 'supplierNo' 214906500ML
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Identifiants chimiques

CAS 6303-21-5
Molecular Formula H3O2P
Molecular Weight (g/mol) 66.00
MDL Number MFCD02183592
InChI Key ACVYVLVWPXVTIT-UHFFFAOYSA-N
Synonym hypophosphite ion, hypophosphorus acid, phosphinic acid, ion 1-, hydroxyphosphanone, oxo-, e5-phosphanol, hydroxidooxidophosphorus, perfluoroalkyl c6-c12 phosphonic acid, diperfluoroalkyl c6-c12 phosphinic acid, hydroxy-, e5-phosphanone, perfluoro-c6-c12 ethyl hydrogen phosphate
PubChem CID 22497
ChEBI CHEBI:36363
IUPAC Name phosphinic acid
SMILES O[PH2]=O

Spécification

Concentration 50wt. % in H2O
Density 1.206g/mL at 25°C
Vapor Pressure 17mmHg (20°C)
Quantity 500 mL
Linear Formula H3PO2
UN Number 3264
Formula Weight 66g/mol
Chemical Name or Material Hypophosphorous Acid Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.