missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies HypoThermosol™ FRS

Product Code. 17149921
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
16 x 10 mL
500 mL Bag
500 mL Bottle
Unit Size:
100mL
500mL
Pack of 16
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17149921 100 mL 100mL
17109931 500 mL Bag 500mL
17139921 16 x 10 mL Pack of 16
17159921 500 mL Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17149921 Supplier STEMCELL Technologies Supplier No. 07935
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Animal component-free, defined hypothermic (2 - 8°C) preservation medium for a range of cell and tissue types

  • HypoThermosol™ FRS is an optimized hypothermic (2 - 8°C) preservation medium that enables improved and extended preservation of cells, tissues, and organs
  • HypoThermosol™ FRS is uniquely formulated to address the molecular-biological response of cells during the hypothermic preservation process
  • HypoThermosol™ FRS includes key ions at concentrations that balance the intracellular state at hypothermic temperatures
  • Additional components include pH buffers, energy substrates, free radical scavengers, and osmotic/oncotic stabilizers

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells; Mesenchymal Stem and Progenitor Cells; Other; Pluripotent Stem Cells
For Use With (Application) Cryopreservation
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.