missing translation for 'onlineSavingsMsg'
Learn More

Hypoxanthine, 99.5%

Product Code. 10530961 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10530961 25 g Glass bottle 25g
10787763 5 g Glass bottle 5g
10331294 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10530961 Supplier Thermo Scientific Acros Supplier No. 122010250
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 68-94-0 | C5H4N4O | 136.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-94-0
Molecular Formula C5H4N4O
Molecular Weight (g/mol) 136.11
MDL Number MFCD00005725
InChI Key FDGQSTZJBFJUBT-UHFFFAOYSA-N
Synonym hypoxanthine, 6-hydroxypurine, 9h-purin-6-ol, sarkin, 6-oxopurine, sarcine, sarkine, 3h-purin-6-ol, 7h-purin-6-ol, purin-6 1h-one
PubChem CID 790
ChEBI CHEBI:17368
SMILES O=C1N=CNC2=C1NC=N2

Specifications

Melting Point >250.0°C
Color Brown to Colorless
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 26, 416
Merck Index 15, 4908
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in diluted alkalies and acids, practically insoluble in most common organic, solvents
IUPAC Name 3,7-dihydropurin-6-one
Transmittance 95% min. (c=0.1, 0.2M NaOH, 430nm) 1cm cell
Formula Weight 136.11
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material Hypoxanthine
Show More Show Less
compliance-icons
Product Identifier
  • Hypoxanthine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.