missing translation for 'onlineSavingsMsg'
Learn More

Hypoxanthine, TRC

Product Code. 30136245
Change view
Click to view available options
Quantity:
25 mg
1 g
5 g
Unit Size:
1g
25mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136245 5 g 5g
17963405 25 mg 25mg
17914375 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136245 Supplier Toronto Research Chemicals Supplier No. TRCH9985005G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-94-0
Molecular Formula C5 H4 N4 O
Molecular Weight (g/mol) 136.11
Synonym 1,7-Dihydro-6H-purin-6-one, Hypoxanthine, Didanosine Related Compound A (USP), Didanosine Imp. A (Ph. Int.), Didanosine Imp. A (EP), 6H-Purin-6-one, 1,9-dihydro-, 6H-Purin-6-one, 1,7-dihydro- (9CI), Hypoxanthine (8CI), 1,9-Dihydro-6H-purin-6-one, 1,9-Dihydro-purin-6-one, 1H,7H-Hypoxanthine, 1H,9H-Hypoxanthine, 1H-Purin-6(9H)-one, 3H-Purin-6-ol, 6-Hydroxy-1H-purine, 6-Hydroxypurine, 6-Oxopurine, 9H-Purin-6(1H)-one, 9H-Purin-6-ol, Hypoxanthine enol, NSC 129419, NSC 14665, Purin-6(1H)-one, Purin-6(3H)-one, Purin-6-ol, Sarcine, Sarkin, Sarkine
IUPAC Name 1,9-dihydropurin-6-one
SMILES O=C1NC=Nc2[nH]cnc12

Specifications

Quantity 5 g
InChI Formula InChI=1S/C5H4N4O/c10-5-3-4(7-1-6-3)8-2-9-5/h1-2H,(H2,6,7,8,9,10)
Formula Weight 136.0385
Percent Purity >95%
Chemical Name or Material Hypoxanthine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.