Learn More
Beckman Coulter Life Sciences Hyrtel™ Tubing
Product Code. 31777236 Shop All Beckman Coulter Life Sciences Products
Change view
Click to view available options
Length (English):
1 ft.
10 ft.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
31777236 1 ft. Each
31777234 10 ft. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31777236 Supplier Beckman Coulter Life Sciences Supplier No. 960011
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Material for flexibility and chemical resistance
  • Part of the MET ONE Facility Monitoring System
  • Tubing for low flow rate particle counters
Specifications
Product Type Sample Tubing
Display Type None
Color Black
Diameter (English) 1/4 in., 3/8 in.
Length (English) 1 ft.