missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyrTrp Cell Dissociation Reagent

Product Code. 17205814
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17205814 100 mL 100mL
17215814 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17205814 Supplier Cytiva Supplier No. SV30209.01
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 21-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Recombinant and animal-derived component-free alternative to traditional porcine trypsin

  • Animal-derived component-free (ADCF)
  • Dye-free
  • Suitable for a variety of applications and cell types
  • Effective cell dissociation
  • Does not impact cell line viability and doubling times
  • For cell-based vaccine production and stem cell/primary cell/gene therapy cell culture processes
  • For use with attachment-dependent cell types MRC-5, Vero, CHO K-1 cell lines grown with or without serum
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20 °C
Product Type Cell Culture Dissociation Reagent
For Use With Attachment-dependent cell types MRC-5, Vero, CHO K-1 cell lines grown with or without serum
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.