Learn More
uvex i-5 Welding Safety Glasses
Product Code. 17767679 Shop All Uvex Products
Change view
Click to view available options
Lens Tint:
Welding Shade 1.7
Welding Shade 3
Welding Shade 5
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Tint Unit Size
17767679 Welding Shade 5 Each
17747679 Welding Shade 1.7 Each
17757679 Welding Shade 3 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17767679 Supplier uvex Supplier No. 9183045
  • ONLINE EXCLUSIVE PRICE
    19.25 €
    17.18 € / Pack of 1
    Save 2.07 €
    10% Off
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Reliable protection thanks to an innovative, modern design: with x-tended eyeshield and sideshield, the Uvex i-5 can be relied on to protect the eyes in many situations.
Sophisticated features ensure superior wearer comfort even during long periods of use.
General features
  • Sporty premium welding safety glasses with a variety of features and customization options
  • Robust polycarbonate lens with proven uvex infradur coating technology – permanently anti-fog and scratch-resistant lens
  • Optional extra frame provides greater protection against particles, dust and dirt
  • Metal-free
  • Frame color: black, green
Protection features
  • Certified in line with EN 166 (personal eye protection) and EN 169 (welding shade)
  • IR protection according to EN 169 with perfect color recognition according to EN 172
  • UV400 – 100% protection against dangerous UVA and UVB radiation to 400 nm
Comfort features
  • X-tended eyeshield: soft-seal frame technology wraps the forehead and provides protection against particle penetration
  • X-tended sideshield offers additional side protection
  • Ergonomically shaped frame with multi-stage adjustable side arms allows a high degree of customization in order to guarantee a good fit on any head or face shape
  • Soft ear pieces for a comfortable, secure hold without creating pressure points
  • Soft, flexible nose pad adapts to the wearer and ensures a comfortable, pressure-free fit
TRUSTED_SUSTAINABILITY
Specifications
Lens Type Infradur Plus
Lens Coating Anti-fog
Description Anti-fog, Resistant to Welding Sparks, Scratch Resistant
Certifications/Compliance (Europe) EN 166, EN 169
Lens Tint Welding Shade 5
Product Type Welding Spectacles
Frame Color Green, Black
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400