missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ I-BRD9

missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
missing translation for 'unitSize'
10mg
50mg
Description
I-BRD9 is a potent and selective BRD9 inhibitor (pIC50 = 7.3). Exhibits >70-fold selectivity for BRD9 over a panel of 34 other bromodomains and >700-fold selectivity over the BET family. Downregulates CLEC1, DUSP6, FES and SAMSN1 genes in Kasumi-1 cells.
Specifications
Specifications
| Chemical Name or Material | 5-Ethyl-4,5-dihydro-4-oxo-N-(tetrahydro-1,1-dioxido-2H-thiopyran-4-yl)-7-[3-(trifluoromethyl)phenyl]thieno[3,2-c]pyridine-2-carboximidamide |
| CAS | 1714146-59-4 |
| Quantity | 10 mg |
| Target | Bromodomain Inhibitors |
| Molecular Formula | C22H22F3N3O3S2 |
| Formula Weight | 497.55 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction