missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uvex i-guard+ Spectacles
Shop All Uvex Products
Click to view available options
Lens Tint:
Clear
Gray
Unit Size:
Each
Description
- Member of the uvex i-range - versatile 3-in-1 safety eyewear range – allows eyewear to be adapted according to workplace hazards
- Large, flat lens delivers unrestricted field of vision
- X-tended sideshield offers additional protection from hazards
- Soft, flexible face seal adapts to the wearer providing both protection and comfort. It also protects against droplets or splashes of liquids and large dust particles (markings 3 and 4)
- Increased impact protection – markings BT
- Robust polycarbonate lens with uvex's proven supravision coating technology – permanent anti-fog and scratch resistant lens
- UV400 – 100 % protection against dangerous UVA and UVB radiation up to 400 nm
- Metal-free
Specifications
Specifications
| Lens Coating | Anti-fog, Scratch-resistant |
| Color | Anthracite, Blue |
| Certifications/Compliance (Europe) | EN 166, EN 170 |
| Lens Tint | Clear |
| Product Type | Anti-fog Goggles |
| Headband Material | Textile |
| Material | Polycarbonate |
| Disposable | Reusable |
| Lens Material | Polycarbonate |
| Frame Material | Plastic |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction