Learn More
uvex i-guard Spectacles
Product Code. 17757669 Shop All Uvex Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Tint Unit Size
17757669 Gray Each
17727669 Clear Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17757669 Supplier uvex Supplier No. 9143282
  • ONLINE EXCLUSIVE PRICE
    17.60 €
    16.01 € / Pack of 1
    Save 1.59 €
    9% Off
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Uvex i-guard is part of the new Uvex i-range – the innovative flexible 3-in-1 safety eyewear line with suitable products for nearly every workplace hazard or environment.
  • Member of the uvex i-range - versatile 3-in-1 safety eyewear range – allows eyewear to be adapted according to workplace hazards
  • Large, flat lens delivers unrestricted field of vision
  • X-tended sideshield offers additional protection from hazards
  • Soft, flexible face seal adapts to the wearer providing both protection and comfort. It also protects against droplets or splashes of liquids and large dust particles (markings 3 and 4)
  • Side arms with multi-incline points allow for a high level of individualization to ensure a good fit for all facial shapes and head widths
  • Ergonomically shaped side arms provide a secure fit without pressure
  • Robust polycarbonate lens with uvex’s proven supravision coating technology – permanent anti-fog and scratch resistant lens
  • UV400 – 100 % protection against dangerous UVA and UVB radiation up to 400 nm
  • Metal-free
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Scratch-resistant
Color Anthracite, Lime
Certifications/Compliance (Europe) EN 166, EN 170
Lens Tint Gray
Product Type Ergonomic Safety Glasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Standard