missing translation for 'onlineSavingsMsg'
Learn More

i-Inositol, MedChemExpress

Product Code. 16471108
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471108 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16471108 Supplier Medchem Express Supplier No. HYB1411
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress i-Inositol is a chemical compound, associated lipids are found in many foods, in particular fruit, especially cantaloupe and oranges.

i-Inositol is a chemical compound, associated lipids are found in many foods, in particular fruit, especially cantaloupe and oranges.

Specifications

Chemical Name or Material i-Inositol
CAS 87-89-8
Color White
Purity Grade Notes Research
Molecular Formula C6H12O6
Quantity 5 g
Synonym myo-Inositol meso-Inositol
Solubility Information H2O : ≥ 100 mg/mL (555.06 mM) ∣DMSO : 10 mg/mL (55.51 mM; Need ultrasonic)
SMILES O[C@H]1[C@H]([C@H]([C@@H]([C@H]([C@@H]1O)O)O)O)O
Molecular Weight (g/mol) 180.16
Formula Weight 180.16
Percent Purity 98.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.