missing translation for 'onlineSavingsMsg'
Learn More

uvex i-lite Planet Spectacles

Product Code. 17639795 Shop All Uvex Products
Change view
Click to view available options
Anti-fog, Chemical-resistant, Scratch-resistant
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17639795 Anti-fog, Chemical-resistant, Scratch-resistant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17639795 Supplier uvex Supplier No. 9143295
  • ONLINE EXCLUSIVE PRICE
    14.50 €
    13.33 € / Pack of 1
    Save 1.17 €
    8% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Versatile 3-in-1 protective eyewear, which allows a flexible choice for many applications.

The uvex i-range is our new flexible 3-in-1 safety eyewear line with the right product for many workplace hazards or environments. The uvex i-range planet variant also sets standards in terms of sustainability: we use recycled and bio-based materials for our innovative safety glasses as part of environmentally friendly and resource-saving production. The properties of our safety glasses (e.g. mechanical strength, UV protection, design and comfort) are not affected in any way. The uvex i-range planet is flexible in use and contributes effectively to environmental protection – true to our motto: protecting planet.

General features

  • Versatile 3-in-1 protective eyewear range, which allows a flexible choice for many applications
  • Large, flat lens design for an unrestricted field of vision
  • Robust polycarbonate lens with proven uvex supravision coating technology – permanently anti-fog and scratch-resistant lens
  • Sustainably produced by the use of bio-based materials
  • Metal-free
  • Frame color: anthracite, turquoise
  • PC lens: clear

Protection features

  • Certified in line with EN 166 (personal eye protection) and EN 170 (UV filters)
  • Marking: W 166 F CE – 2C-1.2 W1 FKN CE
  • UV400 – 100% protection against dangerous UVA and UVB radiation to 400 nm

Comfort features

  • X-tended sideshield offers additional side protection
  • Multi-stage adjustable side arms allow a high degree of customisation in order to guarantee a good fit on any head or face shape
  • Ergonomically shaped side arms for a perfect fit without creating pressure points
TRUSTED_SUSTAINABILITY

Specifications

Lens Type Chemical-resistant
Lens Coating Anti-fog, Scratch-resistant
Certifications/Compliance (Europe) EN 166, EN 170
Lens Tint Clear
Product Type Ergonomic Safety Glasses
Frame Color Anthracite, Teal Green/
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.