missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ N-(1-Pyrene)Iodoacetamide

Product Code. 11599256
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11599256 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11599256 Supplier Molecular Probes™ Supplier No. P29

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

N-(1-pyrene)iodoacetamide

The thiol-reactive N-(1-pyrene)iodoacetamide can be used to create environment-sensitive bioconjugates with this unique fluorophore.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mg
Product Type N-(1-Pyrene) Iodoacetamide
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Thiol
Shipping Condition Room Temperature
Reactive Moiety Alkyl Halide, Iodoacetamide
Label or Dye Pyrene
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.