Learn More
Masterflex™ I/P™ Standard Pump Head End Bell Assembly
Product Code. 15551136 Shop All Masterflex Products
Change view
Click to view available options
Material:
Polycarbonate
Unit Size:
Each
Product Code. Material Unit Size
15551136 Polycarbonate Each
1 options
Product Code. 15551136 Supplier Masterflex™ Supplier No. 0701974
  • ONLINE EXCLUSIVE PRICE
    225.00 €
    210.44 € / Pack of 1
    Save 14.56 €
    6% Off
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Masterflex™ I/P™ Standard Pump Head End Bell Assembly for use with I/P™ Standard Pump Heads
  • Replacement parts to keep your pump head operating even longer
  • Two end bell assemblies are required for a complete head assembly
  • Choose replacement parts suitable for your I/P™ Standard pump head size
  • Suitable for use with I/P™ 82 Standard Pump Head
  • Constructed with durable Polycarbonate (PC) Housing and Cold-Rolled Steel (CRS) Rotor
Specifications
For Use With I/P™ 82 Standard Pump Head
Material Polycarbonate
Tubing Type I/P™ 82 Precision Tubing
Rotor Material Cold-rolled Steel