missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Silencer™ GAPDH Positive Control siRNA, in vivo ready

Product Code. 10117734
Change view
Click to view available options
Quantity:
250 nmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10117734 250 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10117734 Supplier Invitrogen™ Supplier No. 4404025
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Ambion™ Silencer™ GAPDH Positive Control siRNA, In Vivo Ready is ideal for developing and optimizing siRNA experiments.

Ambion™ Silencer™ GAPDH Positive Control siRNA, In Vivo Ready is ideal for developing and optimizing siRNA experiments. The control is validated for use in human cell lines and optimized for mouse and rat. It is highly purified as required for animal use. 250 nmol is provided.

  • Validated siRNA controls for optimizing siRNA experiments
  • Gene-specific control siRNAs provided with scrambled, negative controls
  • Functionally tested in several common cell lines
  • HPLC purified, duplexed, and ready to use

Quality Control:

Silencer™ siRNAs are manufactured in a state-of-the-art facility under the highest quality standards. After HPLC purification and annealing, each siRNA is further purified utilizing a process that removes excess salt via a semipermeable membrane. The result is highly pure siRNA with minimal salt content, suitable for in vivo applications. In vivo siRNAs are then sterile filtered and tested for the presence of endotoxin. At concentrations of 50 μM in deionized water, In Vivo Ready siRNAs contain less than 0.6 mM Na+, less than 2.0 mM K+, and less than 0.1 mM Mg2+. Silencer™ control siRNAs are unmodified 21-mers; they are designed to be used with Silencer™ siRNAs, BLOCK-iT™ siRNAs, and other unmodified 21mer siRNAs. If you are using other siRNA products, such as Ambion™ Silencer™ Select siRNAs or STEALTH RNAi™ siRNAs, we recommend that you use control siRNAs designed for use with those siRNAs for more reliable experimental results.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The siRNA is provided dried down in a single tube, which should be stored at -20°C.
Description The siRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C.
Control Type Gene Knockdown Positive Control
RNAi Type siRNA
For Use With (Application) RNAi
Label or Dye Unconjugated
Product Line Silencer, Ambion
Product Type siRNA
Purity HPLC
Quantity 250 nmol
Shipping Condition Room Temperature
Target GAPDH
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.