missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Silencer™ KIF11 (Eg5) siRNA (human, mouse, rat)

Product Code. 10701937
Click to view available options
Quantity:
5 nmol
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10701937

Brand: Invitrogen™ AM4639

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Silencer™ KIF11 (Eg5) siRNA (human, mouse, rat)

Invitrogen™ Silencer™ KIF11 (Eg5) Positive Control siRNA (Human, Mouse, Rat) is ideal for developing and optimizing siRNA experiments. 5 nmol is provided, along with an additional tube of 2 nmol negative control siRNA.
  • Validated siRNA controls for optimizing siRNA experiments
  • Gene-specific control siRNAs provided with scrambled, negative controls
  • Functionally tested in several common cell lines
  • HPLC purified, duplexed, and ready to use Silencer™ control siRNAs are unmodified 21-mers; they are designed to be used with Silencer™ siRNAs, BLOCK-iT™ siRNAs, and other unmodified 21mer siRNAs. If you are using other siRNA products, such as Invitrogen™ Silencer™ Select siRNAs or STEALTH RNAi™ siRNAs, we recommend that you use control siRNAs designed for use with those siRNAs for more reliable experimental results.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Store at or below -20°C. Do not store in a frost-free freezer.
    Control Type Positive Control
    RNAi Type siRNA
    Species Human, Mouse, Rat
    For Use With (Application) Transfection, RNAi
    Label or Dye Unconjugated
    Product Line Silencer, Ambion
    Product Type siRNA
    Purity HPLC
    Quantity 5 nmol
    Shipping Condition Dry Ice
    Target KIF11
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.