missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Silencer™ Negative Control No. 2 siRNA

Product Code. 10127774
Change view
Click to view available options
Quantity:
40 nmol
5 nmol
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10127774 5 nmol Each
10554255 40 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10127774 Supplier Invitrogen™ Supplier No. AM4613
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Silencer™ Negative Control No. 2 siRNA

Invitrogen™ Silencer™ Negative Control #2 siRNA has no significant sequence similarity to mouse, rat, or human gene sequences. The control has also been tested in cell-based screens and proven to have no significant effect on cell proliferation, viability, or morphology.

  • Nontargeting with limited sequence similarity to known genes
  • Validated for use in human, mouse, and rat cells
  • Functionally proven to have minimal effects on cell proliferation and viability
  • HPLC purified, duplexed, and ready to use
Negative control siRNAs are essential for determining transfection efficiency and to control for the effects of siRNA delivery. In siRNA screening applications, negative control siRNAs are also critical for setting the “hit“ threshold that determines whether an siRNA is considered to have a positive, negative, or neutral effect in a particular assay.

Accessory Product: Silencer™ siRNA Screening Control Panel (SKU#AM4640)
The Silencer™ siRNA Screening Control Panel includes seven negative control siRNAs, and is for those researchers performing siRNA screens that employ hundreds or thousands of siRNAs per experiment. Because negative control siRNAs may have unintended effects in a particular assay, a negative control siRNA should be carefully chosen. The Silencer™ siRNA Screening Control Panel simplifies this process. As an added benefit, a positive control siRNA to human, mouse, and rat KIF11 (Eg5) is included in the panel. Knockdown of KIF11, which encodes a kinesin family motor protein, leads to mitotic arrest. Silencer™ control siRNAs are unmodified 21-mers; they are designed to be used with Silencer™ siRNAs, BLOCK-iT™ siRNAs, and other unmodified 21mer siRNAs. If you are using other siRNA products, such as Invitrogen™ Silencer™ Select siRNAs or STEALTH RNAi™ siRNAs, we recommend that you use control siRNAs designed for use with those siRNAs for more reliable experimental results.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at or below -20°C. Do not store in a frost-free freezer.
Control Type Negative Control
RNAi Type siRNA
Species Human, Mouse, Rat
For Use With (Application) Transfection, RNAi
Label or Dye Unconjugated
Product Line Silencer, Ambion
Product Type siRNA
Purity HPLC
Quantity 5 nmol
Shipping Condition Dry Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.