missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Silencer™ siRNA Construction Kit with manual

Product Code. 10299094
Change view
Click to view available options
Includes:
Kit only
Kit with manual
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
10299094 Kit only Each
10085604 Kit with manual Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10299094 Supplier Invitrogen™ Supplier No. AM1620

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
The Invitrogen™ Silencer™ siRNA Construction Kit is for the synthesis of siRNAs by in vitro transcription of DNA templates and costs less per siRNA than chemical synthesis. The kit includes sufficient reagents to synthesize 15 siRNAs.
  • Cut siRNA preparation time—generate up to 15 siRNAs in less than 24 hours
  • Get hundreds of transfections per reaction
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 2X
    Content And Storage DNA Hybridization Buffer, T7 Promoter Primer, Klenow Reaction Buffer, 10X dNTP, Exo– Klenow, T7 Enzyme Mix, T7 Reaction Buffer, 2X NTP Mix, Digestion Buffer, DNase, RNase, Sense Control DNA, and Antisense Control DNA should be stored at –20°C. siRNA Binding Buffer, siRNA Wash Buffer, Filter Cartridges, and Tubes should be stored at room temperature. Nuclease-free Water may be stored at any temperature.
    Control Type Transcription Control
    Sample Type In vitro Transcribed siRNA
    Includes Kit only
    No. of Reactions 15
    Product Line Silencer, Ambion
    Quantity 15 reactions
    Shipping Condition Dry Ice

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.