missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Silencer™ siRNA Construction Kit with manual

Product Code. p-7180302
Click to view available options
Includes:
Kit only
Kit with manual
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10299094

Brand: Invitrogen™ AM1620

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The Invitrogen™ Silencer™ siRNA Construction Kit is for the synthesis of siRNAs by in vitro transcription of DNA templates and costs less per siRNA than chemical synthesis. The kit includes sufficient reagents to synthesize 15 siRNAs.
  • Cut siRNA preparation time—generate up to 15 siRNAs in less than 24 hours
  • Get hundreds of transfections per reaction
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 2X
    Content And Storage DNA Hybridization Buffer, T7 Promoter Primer, Klenow Reaction Buffer, 10X dNTP, Exo– Klenow, T7 Enzyme Mix, T7 Reaction Buffer, 2X NTP Mix, Digestion Buffer, DNase, RNase, Sense Control DNA, and Antisense Control DNA should be stored at –20°C. siRNA Binding Buffer, siRNA Wash Buffer, Filter Cartridges, and Tubes should be stored at room temperature. Nuclease-free Water may be stored at any temperature.
    Control Type Transcription Control
    Sample Type In vitro Transcribed siRNA
    Includes Kit only
    No. of Reactions 15
    Product Line Silencer, Ambion
    Quantity 15 reactions
    Shipping Condition Dry Ice

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.