missing translation for 'onlineSavingsMsg'
Learn More

Iberin, TRC

Product Code. 17954415
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17954415 5 mg 5mg
17953615 10 mg 10mg
17922815 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17954415 Supplier Toronto Research Chemicals Supplier No. TRCI1249005MG
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 505-44-2
Molecular Formula C5 H9 N O S2
Molecular Weight (g/mol) 163.26
Synonym 1-Isothiocyanato-3-(methylsulfinyl)-propane, 3-Methylsulfinylpropyl Isothiocyanate, NSC 321801;
IUPAC Name 1-isothiocyanato-3-methylsulfinylpropane
SMILES CS(=O)CCCN=C=S

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C5H9NOS2/c1-9(7)4-2-3-6-5-8/h2-4H2,1H3
Formula Weight 163.0126
Chemical Name or Material Iberin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.