missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBlot™ 2 Transfer Stacks, PVDF, regular size

Product Code. 15553267
Change view
Click to view available options
Quantity:
10 Stacks
3 x 10 stacks
Unit Size:
Pack of 10
Pack of 3
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15553267 3 x 10 stacks Pack of 3
15249296 10 Stacks Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15553267 Supplier Invitrogen™ Supplier No. IB24001X3
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

iBlot 2 Transfer Stacks are used to transfer proteins using the iBlot 2 Gel Transfer Device. iBlot 2 Transfer Stacks are consumable stacks with an integrated pre-activated PVDF transfer membrane for dry blotting of proteins.

iBlot 2 Transfer Stacks are used to transfer proteins using the iBlot 2 Gel Transfer Device. iBlot 2 Transfer Stacks are consumable stacks with an integrated pre-activated PVDF transfer membrane for dry blotting of proteins. Each iBlot 2 Transfer Stack contains a copper-coated electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins. This regular-size stack (13 x 8.3 cm) can be used with midi-size gels or two mini-size gels or E-PAGE 48 or 96 gels.

PVDF membrane
The PVDF membrane (0.2-μm pore size, low fluorescence) in this stack has higher binding capacity than nitrocellulose. It is pre-activated and ready for use without any pretreatment with alcohols. The membrane is compatible with commonly used detection methods such as staining, immunodetection, fluorescence, and radiolabeling. The proteins bind to the membrane due to hydrophobic interactions. The protein-binding capacity is 240 μg/cm2.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 3 x 10 stacks
Format Transfer Stack
Material PVDF
Pore Size 0.2 μm
Content And Storage Store at room temperature.
Dimensions (LxW) 13.5 x 8 cm
Product Line iBlot

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.