missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBlot™ 3 Electrode Replacement Kit

Product Code. 31662775
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31662775 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31662775 Supplier Invitrogen™ Supplier No. A56427

Please to purchase this item. Need a web account? Register with us today!

The iBlot 3 Electrode Replacement Kit contains four electrical contacts, four white rubber seals, and eight screws for the iBlot 3 Western Blot Transfer Device.

The iBlot 3 Electrode Replacement Kit contains four electrical contacts, four white rubber seals, and eight screws for the iBlot 3 Western Blot Transfer Device. Please refer to the iBlot 3 Western Blot Transfer System User Guide for instructions on how to replace the electrical contacts.

About the iBlot 3 Western Blot Transfer Device
The iBlot 3 Western Blot Transfer Device (Cat. No IB31001) is a dry transfer device that performs western blotting transfer efficiently and reliably in as few as three minutes and without the need for liquid buffers. The iBlot 3 system is compatible with both nitrocellulose and polyvinylidene difluoride (PVDF) membranes and offers equivalent or better performance compared with traditional wet-transfer methods in a fraction of the time. The iBlot 3 Western Blot Transfer Device is an integral part of the iBlot dry-blotting system, which consists of the transfer device and consumable transfer stacks that contain the required buffers and transfer membrane (nitrocellulose or PVDF).

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Four electrical contacts
• Four white rubber seals
• Eight screws

Store at room temperature.

For Use With (Equipment) iBlot 3
Gel Compatibility Mini- or Midi-sized Bis-Tris Gels, Tricine Gels, Tris-Acetate Gels, and Tris-Glycine Gels
Membrane Compatibility Nitrocellulose, PVDF
Mode of Transfer Dry
Product Line iBlot 3
Type Electrode Replacement Kit
Quantity 1 kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.