missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies IBMX

Product Code. 17120122
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17120122 100 mg 100mg
17130122 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17120122 Supplier STEMCELL Technologies Supplier No. 72762
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

cAMP pathway activator; Inhibits cyclic nucleotide phosphodiesterases

  • IBMX is an inhibitor of cyclic nucleotide phosphodiesterases (PDEs; IC50 = 19, 50, 18, 13, 32, 7, and 50 μM for PDE1, PDE2, PDE3, PDE4, PDE5, PDE7, and PDE11, respectively)
  • By inhibiting PDEs, IBMX increases cellular cAMP and cGMP levels, activating cyclic-nucleotide-regulated protein kinases

Specifications

CAS 28822-58-4
Synonym 1-Methyl-3-Isobutylxanthine; Isobutylmethylxanthin; Isobutyl methylxanthine; NSC 165960
Quantity 100 mg
Molecular Formula C10H14N4O2
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.