missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ IBMX

Product Code. 18064282
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18064282 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18064282 Supplier Tocris Bioscience™ Supplier No. 2845/50
  • ONLINE EXCLUSIVE PRICE
    146.00 €
    137.55 € / 50mg
    Save 8.45 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PDE inhibitor (non-selective)

Phosphodiesterase inhibitor (IC50 values are 13, 18, 19, 32 and 50 μM for PDE4, PDE3, PDE1, PDE5 and PDE2 respectively). Suppresses α-adrenoceptor-mediated 5-HT release from neuroendocrine epithelial cells (IC50 = 1.3 μM). Facilitates differentiation of neural progenitor cells in vitro.
TRUSTED_SUSTAINABILITY

Specifications

CAS 28822-58-4
Quantity 50 mg
Molecular Formula C10H14N4O2
Formula Weight 222.24
Purity >99%
Molecular Weight (g/mol) 222.25
Chemical Name or Material IBMX
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.