missing translation for 'onlineSavingsMsg'
Learn More

Ibotenic acid, MedChemExpress

Product Code. p-200098054
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16479278

Brand: Medchem Express HYN2311

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ibotenic acid has agonist activity at both the N-methyl-D-aspartate (NMDA) and trans-ACPD or metabolotropic quisqualate (Qm) receptor sites.

Ibotenic acid has agonist activity at both the N-methyl-D-aspartate (NMDA) and trans-ACPD or metabolotropic quisqualate (Qm) receptor sites.

Specifications

Chemical Name or Material Ibotenic acid
CAS 2552-55-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C5H6N2O4
Quantity 10 mg
Synonym (RS)-Ibotenic acid DL-Ibotenic acid
Solubility Information H2O : 5 mg/mL (31.62 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (31.62 mM; Need ultrasonic)
SMILES O=C(O)C(N)C(ON1)=CC1=O
Molecular Weight (g/mol) 158.11
Formula Weight 158.11
Percent Purity 98.27%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.