missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ibotenic acid

Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
NMDA and metabotropic glutamate receptor agonist.
Specifications
Specifications
| CAS | 2552-55-8 |
| Quantity | 5 mg |
| Molecular Formula | C5H6N2O4 |
| Formula Weight | 158.11 |
| Purity | >98% |
| Molecular Weight (g/mol) | 158.113 |
| Chemical Name or Material | Ibotenic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction