missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBright™ Imaging System White Transilluminator Screen

Product Code. 15716285
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15716285 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15716285 Supplier Invitrogen™ Supplier No. A33828
  • ONLINE EXCLUSIVE PRICE
    183.00 €
    148.23 € / Pack of 1
    Save 34.77 €
    19% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Replacement white transilluminator screen for the iBright imaging systems

  • This is a replacement white transilluminator screen for the iBright imaging systems (CL1000, CL1500, FL1000, and FL1500).
  • The white transilluminator screen provides a contrasting background when imaging protein gels stained with colorimetric stains such as coomassie and silver stains.
  • This transilluminator screen is automatically detected by the iBright imaging system and is required to provide the desired image black/white orientation and band identification with colorimetric stained gels.
TRUSTED_SUSTAINABILITY

Specifications

Product Line iBright
For Use With (Application) Chemiluminescent Western Blot Imaging, DNA Gel Imaging, Protein Gel Imaging
Product Type Imaging System
Quantity 1 Ea.
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.