Learn More
Invitrogen™ iBright™ SAE Software for 21 CFR Part 11, single license
Shop All Thermo Scientific ProductsDescription
- Authentication of user login information
- Ability to set password guidelines and expiry dates
- Lockout access and re-authentication requirements after a period of inactivity
- System lockout upon unauthorized attempts to access software
- Ability of administrator to define user roles and permissions
- Verification of each user's permissions to perform predefined activities
- Identification of data tampering
- Generation of audit trail log
- Tracking of changes and committed actions to user roles, account management, and system settings
- Ability of administrator to determine functions required to input reason for commit
- Administrator determination of e-signature requirements
- Ability to review records and sign electronically
Specifications
Specifications
| For Use With (Equipment) | iBright Imaging System |
| Product Line | iBright |
| Type | Software License |
| Quantity | 1 license |
| Software Type | Software License |