missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBright™ System Tray Adapter for E-Gel™ Agarose Gels

Product Code. 17896511
Change view
Click to view available options
Product Type:
E-Gel Tray Adapter, 11-/22-well
E-Gel Tray Adapter, 48-/96-well
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
17896511 E-Gel Tray Adapter, 48-/96-well Each
17806521 E-Gel Tray Adapter, 11-/22-well Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17896511 Supplier Invitrogen™ Supplier No. A56599
  • ONLINE EXCLUSIVE PRICE
    176.00 €
    142.56 € / Pack of 1
    Save 33.44 €
    19% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Imaging accessories designed to optimize the image capture of Invitrogen E-Gel Agarose Gels.

Invitrogen iBright Tray Adapters for E-Gel Agarose Gels are imaging accessories designed to optimize the image capture of Invitrogen E-Gel Agarose Gels.

Features of Invitrogen iBright Tray Adapters for E-Gel Agarose Gels include:
• The bottom component centers the E-Gel agarose gels on the iBright transilluminator box facilitating optimized zoom
• The top component covers labels and bar codes on the E-Gel cassette to reduce stray light during image capture and improve sample signal optimization
• Compatible with all iBright models (CL750, CL1000, CL1500, FL1000, and FL1500)
• Compatible with 11- to 96-well E-Gel agarose (Cat. No. A56600 for 11-/22-well, Cat. No. A56599 for 48-/96-well)

TRUSTED_SUSTAINABILITY

Specifications

Product Line iBright
For Use With (Application) Chemiluminescent Western Blot Imaging, DNA Gel Imaging, Protein Gel Imaging
Product Type E-Gel Tray Adapter, 48-/96-well
Quantity 1 Adaptor
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.