missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ iBu-dG Phosphoramidite, standard grade, 20-400 finish bottle

Product Code. 17967172 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17967172 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17967172 Supplier Thermo Scientific™ Supplier No. 27173411
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

iBu-dG CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyguanosine (dG) with isobutyryl (iBu) base protection, for use in oligonucleotide synthesis.

iBu-dG CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyguanosine (dG) with isobutyryl (iBu) base protection, for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups designed specifically to protect the DNA during the chemical synthesis process, and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. The 'Standard Grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 30 mL Amber bottle with a 20-400 Finish neck style, which is compatible with most Expedite Systems of oligonucleotide synthesizers.

Features:
Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-deoxyguanosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
Molecular formula: C44H54N7O8P
Base: dG
Base Protection: iBu
Formula weight: 839.92 g/mol
CAS number: 93183-15-4
HPLC Purity: ≥98.0%
31P NMR Purity: ≥98.0%

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Expedite Synthesizers
Product Type iBu-dG CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Deoxyguanosine (dG)
Shipping Condition Room Temperature
Purity 98%
Quantity 2 g
Reagent Type Phosphoramidite
Grade Standard
Nucleic Acid Type DNA
Form Powder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.