Learn More
Thermo Scientific™ TheraPure™ iBu-dG Phosphoramidite, 20-400 finish bottle
Shop All Thermo Scientific ProductsDescription
iBu-dG CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyguanosine (dG) with isobutyryl (iBu) base protection, for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups designed specifically to protect the DNA during the chemical synthesis process, and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. The TheraPure trademark denotes a highest level of phosphoramidite purity, developed with the highest stringency of of high definiteion analytics and tightly controlled impurity profiles. Ideal for any therapeutic application. This product is supplied in a 60 mL Amber bottle with a 20-400 Finish neck style, which is compatible with most Expedite Systems of oligonucleotide synthesizers.
Features:
• Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-deoxyguanosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula: C44H54N7O8P
• Base: dG
• Base Protection: iBu
• Formula weight: 839.92 g/mol
• CAS number: 93183-15-4
• HPLC Purity: ≥99%; M-11 Impurity ≤0.01%; Single critical impurity ≤0.3%; Presence of dA, dC, or T impurity ≤0.03%
• 31P NMR Purity: ≥99%; Sum of non-primary peaks at 140-152 ppm: ≤0.3%; No peak(s) above 170 ppm; HMT impurity (∼146 ppm): ≤0.10%
Specifications
Specifications
| For Use With (Equipment) | Expedite Synthesizers |
| Product Type | iBu-dG CEP |
| Content And Storage | Store in freezer (at or below -20°C). |
| Nucleoside Type | Deoxyguanosine (dG) |
| Shipping Condition | Room Temperature |
| Purity | 99% |
| Quantity | 2 g |
| Reagent Type | Phosphoramidite |
| Grade | TheraPure™ |
| Nucleic Acid Type | DNA |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.