missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ iBu-rG Phosphoramidite, standard grade, serum vial bottle

Product Code. 17997182 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17997182 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Code produit 17997182 Fournisseur Thermo Scientific™ Code fournisseur 27140607
 Restricted Item
In Stock
Ajouter au panier
Ajouter au panier
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

iBu-rG CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with isobutyryl (iBu) base protection, for use in oligonucleotide synthesis.

iBu-rG CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with isobutyryl (iBu) base protection, for use in oligonucleotide synthesis. RNA phosphoramidites contain a 2'-OH protecting group, tBDMS (tert-butyl dimethylsilyl). Each base has unique protecting groups designed specifically to protect the RNA during the chemical synthesis process, and produce the highest amount of full length product. Alternate base protection options are also available to allow for faster deprotection and longer oligonucleotide synthesis. The 'Standard Grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 60 mL Amber bottle with a Serum Vial neck style, which is compatible with most ABI Systems|AKTA Systems of oligonucleotide synthesizers.

Features:
Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-O-TBDMSi-guanosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
Molecular formula: C50H68N7O9PSi
Base: rG
Base Protection: iBu
Formula weight: 970.1 g/mol
CAS number: 147201-04-5
HPLC Purity: ≥98.0%
31P NMR Purity: ≥98.0%; Sum of non-primary peaks in 140-152 ppm range: ≤0.5%

TRUSTED_SUSTAINABILITY

Spécification

For Use With (Equipment) AKTA Synthesizers, Applied Biosystems Synthesizers
Product Type iBu-rG CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Guanosine (G)
Shipping Condition Room Temperature
Purity 98%
Quantity 2 g
Reagent Type Phosphoramidite
Grade Standard
Nucleic Acid Type RNA
Form Powder
Afficher plus Afficher moins

For Research Use Only. Not for use in diagnostic procedures.

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.