missing translation for 'onlineSavingsMsg'
Learn More

Ibuprofen Acyl-Beta-D-glucuronide (mixture of diastereomers), TRC

Product Code. 30004628
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004628 5 mg 5mg
30009442 10 mg 10mg
30008275 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004628 Supplier Toronto Research Chemicals Supplier No. TRCI1400305MG
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115075-59-7
Molecular Formula C19 H26 O8
Molecular Weight (g/mol) 382.4
Synonym Ibuprofen Acyl-Beta-D-Glucuronide
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[2-[4-(2-methylpropyl)phenyl]propanoyloxy]oxane-2-carboxylic acid
SMILES CC(C)Cc1ccc(cc1)C(C)C(=O)O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C19H26O8/c1-9(2)8-11-4-6-12(7-5-11)10(3)18(25)27-19-15(22)13(20)14(21)16(26-19)17(23)24/h4-7,9-10,13-16,19-22H,8H2,1-3H3,(H,23,24)/t10?,13-,14-,15+,16-,19-/m0/s1
Formula Weight 382.1628
Chemical Name or Material Ibuprofen Acyl-beta-D-glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.