missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Ibutilide hemifumarate

Product Code. 30001488
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001488 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001488 Supplier R&D Systems™ Supplier No. 3908/50
  • ONLINE EXCLUSIVE PRICE
    231.22 €
    207.90 € / 50mg
    Save 23.32 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Blocks IKr, hERG and L-type Ca2+ channels; class III antiarrhythmic

Class III antiarrhythmic. Blocks the rapid component of delayed rectifier K+ current (IKr); also blocks hERG channels and L-type Ca2+ channels. Shown to activate a slow inward Na+ current in guinea pig ventricular cells. Used to terminate atrial fibrillation and atrial flutter; can prolong QT.
TRUSTED_SUSTAINABILITY

Specifications

CAS 122647-32-9
Quantity 50 mg
Product Line Tocris
Molecular Formula C44H76N4O10S2
Formula Weight 442.61
Purity >98%
Molecular Weight (g/mol) 885.23
Chemical Name or Material Ibutilide hemifumarate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.