Learn More
BINDER™ ICH Light Module
Product Code. p-200104333 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB PRO 720, KBF/KBF PRO 720
KB PRO/KBF/KBF PRO 260/470
Unit Size:
Each
Product Code. 30238258
Brand: BINDER™ 80122441
Only null left
Add to basket
Add to basket
ICH-Q1B light module
Module consists of:
  • VIS cassette
  • UVA cassette
  • Independent power supply
Specifications
For Use With (Equipment) KB PRO/KBF/KBF PRO 260/470
Material Glass
Type Lighting