missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ ICI 182,780

Product Code. 30001177
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001177 10 mg 10mg
18742564 1 mg 1mg
18742654 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30001177 Supplier R&D Systems™ Supplier No. 1047/10
  • ONLINE EXCLUSIVE PRICE
    347.86 €
    311.85 € / 10mg
    Save 36.01 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Estrogen receptor antagonist

ICI 182,780 is a high affinity estrogen receptor antagonist (IC50 = 0.29 nM), devoid of any partial agonism both in vitro and in vivo. ICI 182,780 is a high affinity agonist at the membrane estrogen receptor GPER. ICI 182,780 inhibits MCF-7 human breast cancer cell growth in vitro and shows antitumor activity in mice models. ICI 182,780 penetrates the blood-brain-barrier and has neuronal functional effects in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129453-61-8
Molecular Formula C32H47F5O3S

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 606.77
Purity >99%
Molecular Weight (g/mol) 606.777
Chemical Name or Material ICI 182, 780
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.