missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ ICI 204,448 hydrochloride
Description
ICI 204,448 hydrochloride is a peripherally acting κ agonist.
Specifications
Specifications
| Chemical Name or Material | (RS)-[3-[1-[[(3,4-Dichlorophenyl)acetyl]methylamino]-2-(1-pyrrolidinyl)ethyl]phenoxy]acetic acid hydrochloride |
| CAS | 121264-04-8 |
| Assay Percent Range | >98% |
| Quantity | 50 mg |
| Solubility | Soluble to 100mM in DMSO |
| Product Line | Tocris |
| Molecular Formula | C23H26Cl2N2O4.HCl |
| Formula Weight | Observed MW: 501.84 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?