missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ CO2 Incubator ICO

Product Code. 15516425 Shop All Memmert Products
Change view
Click to view available options
Capacity (Metric):
56 L
107 L
156 L
241 L
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15516425 56 L Each
15682244 107 L Each
15578675 156 L Each
15692244 241 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 15516425 Supplier Memmert™ Supplier No. ICO50
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.

Ideal for Incubation with CO2, Biosyntesis and Cell Incubation in the Laboratory.

Sterile, safe and reliable – these three words describe the Memmert CO2 incubator ICO for industrial use.

  • Model sizes: 50, 105, 150, 240
  • Temperature adjustment range: +18 °C to +50 °C
  • CO2 adjustment range: 0 to 20 %
  • Humidity adjustment ranges: 40 to 97 % rh (with option K7)
  • O2 adjustment ranges: 1 to 20 % (with option T6)
  • 1 model variant: TwinDISPLAY (intuitive control)
  • AtmoCONTROL software on a USB stick for programming, managing and transferring programmes via Ethernet interface or USB port
  • Perfect distribution of temperature, humidity and CO2
  • Long-time stable and repetitively accurate
  • The interior chamber, including all installations and sensors, can be sterilised at +180 °C in a 60-minute programme
  • Visual and acoustic alarm as well as alarm notification to mobile phones if the individually adjustable temperature, CO2, O2, or humidity ranges are exceeded
  • Battery-buffered ControlCOCKPIT: The operating display, logging and CO2 control are fully functional even during a power failure
  • Fail-safe, FDA-compliant logging of temperature, CO2, O2 humidity, open door time data
  • Adjustable parameters: temperature (Celsius or Fahrenheit), CO2, programme time, time zones, summertime/wintertime

Active humidity control ensures short recovery times after the door has been opened and minimises vaporisation in the interior. Together with the heating of the interior from all six sides and the heated inner glass door, it offers maximum protection for cell and tissue cultures and avoids the dangerous formation of condensation. The turbulence-free chamber ventilation ensures a constant and uniform atmosphere.

Specifications

Alarm Types Optic and acoustic
Capacity (Metric) Shelf 15 kg
Controller Type TwinDISPLAY: adaptive multifunctional digital PID-microprocessor controller with 2 high-definition TFT-colour displays
Capacity (Metric) 56 L
Chamber Material Stainless Steel
CO2 Concentration Range 0 to 20%
CO2 Sensor Technology IR Sensor
Material Textured Stainless Steel, Rear Zinc-plated Steel
No. of Shelves 1
Oxygen Control 1 to 20%
Relative Humidity 40 to 97%
Temperature Range (Metric) 18°C to 50°C
Type CO2 Incubator
Disinfection Time 60 min.
Display 2 high-definition TFT-colour displays (TwinDISPLAY)
Shelf Material Perforated stainless steel
Timer Digital backwards counter with target time setting, adjustable from 1 minute to 99 days
Dimensions (D x W x H) Interior 330 x 400 x 425 mm
Dimensions (L x W x H) 521 x 559 x 791 mm
Shipping Weight (Metric) 74 kg
Weight (Metric) 55 kg
Electrical Requirements 115/230 V, 50/60 Hz
Voltage 115/230 V
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.